1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%
1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%
1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%
1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%
1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%
Thermo Scientific Chemicals

1,2-Bis(2-aminophenoxy)-ethane-N,N,N',N'-tetraacetic acid, tetrasodium salt hydrate, 95%

CAS: 336624-09-0 | C22H20N2Na4O10 | 564.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 401650010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS336624-09-0
IUPAC Nametetrasodium 2-{[2-(2-{2-[bis(carboxylatomethyl)amino]phenoxy}ethoxy)phenyl](carboxylatomethyl)amino}acetate
Molecular FormulaC22H20N2Na4O10
InChI KeyZWSMLJACYSGFKD-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CN(CC([O-])=O)C1=C(OCCOC2=C(C=CC=C2)N(CC([O-])=O)CC([O-])=O)C=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)White to beige
Infrared spectrumConforms
Titration Complexometric>=94.0 %
Water=<12 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chelation/Complexation reagent
RUO – Research Use Only