Benzamidine hydrochloride, 99%
Benzamidine hydrochloride, 99%
Benzamidine hydrochloride, 99%
Thermo Scientific Chemicals

Benzamidine hydrochloride, 99%

CAS: 1670-14- | C7H9ClN2 | 156.61 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 401790050
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.40
Save 11.75 (15%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
65.65
Special offer
Online exclusive
Ends: 30-Jun-2026
77.40
Save 11.75 (15%)
Each
Chemical Identifiers
CAS1670-14-0
IUPAC Namehydrogen benzenecarboximidamide chloride
Molecular FormulaC7H9ClN2
InChI KeyLZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES[H+].[Cl-].NC(=N)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=98.5 %
Water=<1.0 % (K.F.)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only