Can I use the TaqMan β-actin Detection Reagents (Cat. No. 401846) as an endogenous control for my gene expression studies?
The TaqMan β-actin Detection Reagents were designed to amplify genomic DNA, and thus are not recommended for use as an endogenous control for gene expression studies. However, we do offer other TaqMan assay reagents that were specifically designed for that application. Two examples are: Human ACTB (Beta Actin) Endogenous Control, Cat. No. 4310881E (TAMRA probe) or Cat. No. 4326315E (MGB probe).
There are many factors to consider when validating a specific endogenous control. For additional information on designing gene expression assays and controls, please refer to the following documents: "Protocol: TaqMan Human Endogenous Control Plate", and User Bulletin #2: "Relative Quantitation of Gene Expression". You can find copies of these documents on our website by entering the titles in the main Search field.
There are many factors to consider when validating a specific endogenous control. For additional information on designing gene expression assays and controls, please refer to the following documents: "Protocol: TaqMan Human Endogenous Control Plate", and User Bulletin #2: "Relative Quantitation of Gene Expression". You can find copies of these documents on our website by entering the titles in the main Search field.