ProSorb™ Inserts, refill pack - FAQs
ProSorb™ Inserts, refill pack - FAQs
View additional product information for ProSorb™ Inserts, refill pack - FAQs (402052)
1 product FAQs found
Do you have any recommendations for sequencing of protein samples containing detergents, salts, free amines, and other components which are known to interfere?
Use Prosorb Starter Kit, P/N 402139, which enables the concentration and clean up of proteins or peptides, and application onto a PVDF membrane support.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.