Bismuth(III) nitrate pentahydrate, 98%, for analysis
Bismuth(III) nitrate pentahydrate, 98%, for analysis
Bismuth(III) nitrate pentahydrate, 98%, for analysis
Thermo Scientific Chemicals

Bismuth(III) nitrate pentahydrate, 98%, for analysis

CAS: 10035-06-0 | BiH10N3O14 | 485.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 402610025
Price (EUR)
408,00
Each
Quantity:
2.5 kg
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
408,00
Each
Chemical Identifiers
CAS10035-06-0
IUPAC Namebismuth(3+) pentahydrate trinitrate
Molecular FormulaBiH10N3O14
InChI KeyFBXVOTBTGXARNA-UHFFFAOYSA-N
SMILESO.O.O.O.O.[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration Complexometric>=97.5 %
Insoluble matter=<0.005 % (in HNO3)
Arsenic (As)=<10 ppm
Copper (Cu)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only