Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol
Boron trifluoride, 12% (1.5M) in methanol
Thermo Scientific Chemicals

Boron trifluoride, 12% (1.5M) in methanol

Boron trifluoride, 12% (1.5M) in methanol, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 65.0 deg.C, CAS: 373-57-9, 67-56-1, Amber to Colorless, Melting Point: -98.0 deg.C, Molecular Formula: BF3, Molecular Weight: 67.81, Percent Purity: 10 to 15% (w/v BF3) | CAS: 373-57-9 | BF3 | 67.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 402765000
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
92.65
Online Exclusive
103.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS373-57-9
IUPAC Nametrifluoroborane
Molecular FormulaBF3
InChI KeyWTEOIRVLGSZEPR-UHFFFAOYSA-N
SMILESFB(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Solution
Appearance (Color)Colorless to amber
Titration with NaOH10 to 15 % w/v
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Acylation reagent, Lewis acid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only