TFA Aminolink Phosphoramidite, 0.25 g - FAQs
TFA Aminolink Phosphoramidite, 0.25 g - FAQs
View additional product information for TFA Aminolink Phosphoramidite, 0.25 g - FAQs (402872)
1 product FAQs found
Does sodium azide interfere with AminoLink Support coupling?
No, the gel is stored in sodium azide with no loss in activity. Azide is not a primary amine and is rather unreactive.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.