PRTCL,TAQMAN GOLD RT-PCR KIT - FAQs
PRTCL,TAQMAN GOLD RT-PCR KIT - FAQs
View additional product information for PRTCL,TAQMAN GOLD RT-PCR KIT - FAQs (402876)
1 product FAQs found
Where can I find a protocol describing the use of the TaqMan human GAPDH Control Reagents (Cat. No. 402869)?
The protocol for the use of the TaqMan Human GAPDH Control Reagents is provided within the protocol for the TaqMan Gold RT-PCR Kit. A copy of the document can be found on the product page for Catalog No. 402869.