2-Bromoethyl methyl ether, 95%
2-Bromoethyl methyl ether, 95%
2-Bromoethyl methyl ether, 95%
Thermo Scientific Chemicals

2-Bromoethyl methyl ether, 95%

CAS: 6482-24-2 | C3H7BrO | 138.99 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 402980250
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.60
Save 9.95
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
60.65
Special offer
Online exclusive
Ends: 28-Dec-2026
70.60
Save 9.95
Each
Chemical Identifiers
CAS6482-24-2
IUPAC Name1-bromo-2-methoxyethane
Molecular FormulaC3H7BrO
InChI KeyYZUPZGFPHUVJKC-UHFFFAOYSA-N
SMILESCOCCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
AppearanceMay darken on storage
Infrared spectrumConforms
GC>=94 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only