Applied Biosystems™

NED™ Matrix Standard

Catalog number:  402996

 Related applications:

Genotyping & Genomic Profiling

| Sanger Sequencing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The NED™ Matrix Standard can be used to analyze 6-FAM™, VIC®, NED™ and ROX™-labeled DNA fragments in fragment analysis applications. The standard can be used on the Applied Biosystems® 310 Genetic Analyzers. The Data Collection Software for these instruments uses the multicomponent matrix to automatically analyze the four different colored fluorescent dye-labeled samples in a single capillary.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): 310 Genetic Analyzer
Label or Dye: NED
Product Line: NED™
Product Size: 1 tube
Shipping Condition: Wet Ice

Contents & storage

Contains 1 tube of NED Matrix Standard. Store at 2-8°C. (Do not freeze).

Documents

Manuals & protocols