See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
▸
Shop All Products
▸
Incubators
▸
Incubator Accessories
▸
Shelves for Precision™ Standard Incubators
Actual product may vary
Thermo Scientific™
Shelves for Precision™ Standard Incubators
Increase the flexibility of Precision Standard Incubators with these additional shelves.
Have Questions?
Change view
Catalog Number
Size
403-8Q
5.1 cu. ft. Incubator
203-8Q
1.3 cu. ft. Incubator
2 Options
Catalog number
403-8Q
Price (EUR)
-
Request A Quote
Size:
5.1 cu. ft. Incubator
Benefit from Thermo Scientific™ Precision™ Standard Incubator Shelves, which increase the flexibility of Precision Standard Incubators.
Specifications
Description
Shelf Kit
For Use With (Equipment)
5.1 cu. ft. Precision Standard Incubator
Lighting
LED
Size
5.1 cu. ft. Incubator
Type
Shelf