MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps - FAQs
MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps - FAQs
View additional product information for MicroAmp™ Optical 96-Well Reaction Plate with Barcode & Optical Caps - FAQs (403012)
2 product FAQs found
Can I place a standard MicroAmp 96-Well Optical Reaction Plate onto a 7500 Fast Real-Time PCR System?
No. The plate does not fit and will result in the crushing of the wells and possible contamination of the block due to leaking of the well contents.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.
Which well plates do you recommend using with the QuantStudio 5 Real-Time PCR System?
Please see the following link to our qPCR and PCR Plastics Selection Tool to narrow down the list of products compatible with the QuantStudio 5 Real-Time PCR System: https://www.thermofisher.com/de/en/home/life-science/pcr/pcr-plastics/plastics-selection-guide.html
- For Manufacturer, Select "Thermo Fisher Scientific"
- For Model, Select "Applied Biosystems QuantStudio 5"
- Select the appropriate block format for your qPCR instrument
- Click "Display Results"
To the left of the results list, you can filter the results further by various parameters (i.e., Consumable Type, Number of Wells, etc.)