Product Image
Applied Biosystems™

Control DNA (from CEPH Individual 1347-02)

The Control DNA can be used with Linkage Mapping Sets MD10, HD5, and LD20. • Functions as a control templateRead more
Have Questions?
Catalog NumberQuantity
403062180 μL
Catalog number 403062
Price (USD)
415.00
Each
Quantity:
180 μL
Price (USD)
415.00
Each
The Control DNA can be used with Linkage Mapping Sets MD10, HD5, and LD20.

• Functions as a control template to monitor PCR amplification.
• Serves as sizing reference for controlling gel-to-gel or capillary-to-capillary variation.
• Aids in the comparison of alleles sizes from different gels (allele binning).
• Enables correlation with allele frequency data from external sources such as others databases and laboratories.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Sequencing
IncludesControl Template(s)
No. of Reactions150
Product TypeControl DNA
Quantity180 μL
Shipping ConditionDry Ice
SpeciesHuman
FormatTube
Unit SizeEach
Contents & Storage
This product contains control DNA in Tris buffer, pH 8.0, and EDTA.