Product Image
Applied Biosystems™

6-FAM™ Dye Phosphoramidite

Contains 45 mg 6-FAM™ Dye Phosphoramidite.6-FAM™ Dye Phosphoramidite is used for the 5’ attachment of FAM™ dye during oligonucleotide synthesis.Read more
Have Questions?
Catalog NumberQuantity
40316945 mg
Catalog number 403169
Price (USD)
524.00
Each
Quantity:
45 mg
Price (USD)
524.00
Each
Contains 45 mg 6-FAM™ Dye Phosphoramidite.

6-FAM™ Dye Phosphoramidite is used for the 5’ attachment of FAM™ dye during oligonucleotide synthesis. 6-FAM™ Dye Phosphoramidite is an optically pure isomer (not a mixture).

The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.

• Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers
• Designed and optimized for use with ABI™ synthesizers, guaranteeing reliable nucleic acid synthesis
• We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis

On-instrument labeling typically saves hours of post-synthesis manual processing
Fluorescent label 6-FAM™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. 6-FAM™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)ABI Synthesizers, Guaranteeing Reliable Nucleic Acid Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormatPowder
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product Type6-FAM™ Dye CEP
Quantity45 mg
Reagent TypePhosphoramidite
Shipping ConditionDry Ice
Product LineFAM
Unit SizeEach
Contents & Storage
This reagent should be stored dessicated at -20°C.