Contains 55 mg HEX™ Dye Phosphoramidite.HEX™ Dye Phosphoramidite is used for the 5’ attachment of HEX™ dye during oligonucleotide synthesis.TheRead more
Have Questions?
Change view
Catalog Number
Quantity
Product Type
403170
55 mg
HEX Dye CEP
401526
105 mg
HEX Dye CEP
2 Options
Catalog number 403170
Price (USD)
524.00
Each
Quantity:
55 mg
Product Type:
HEX Dye CEP
Price (USD)
524.00
Each
Contains 55 mg HEX™ Dye Phosphoramidite.
HEX™ Dye Phosphoramidite is used for the 5’ attachment of HEX™ dye during oligonucleotide synthesis.
The label is treated as if it were simply another base: It is inserted at the appropriate place in the sequence and incorporated automatically.
• Specialty phosphoramidites allow online addition of the label during synthesis, saving time in production of labeled probes and primers • Designed and optimized for use with ABI™ synthesizers, guaranteeing reliable nucleic acid synthesis • We offer a range of specialty phosphoramites and linkers to give you the flexibility demanded in oligonucleotide synthesis
On-instrument labeling typically saves hours of post-synthesis manual processing Fluorescent label HEX™ is a single-isomer fluorescein dye designed for detection and analysis of labeled PCR products on ABI PRISM™ genetic analysis systems. HEX™ is equally useful in applications historically employing isotopic labels. As such, it eliminates the hazard associated with handling, storage, and disposal of radioactive materials.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection MethodFluorescence
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormPowder
FormatPowder
Nucleic Acid TypeNone (Modifier)
Nucleoside TypeNone (Modifier)
Product TypeHEX Dye CEP
Quantity55 mg
Reagent TypePhosphoramidite
Sample TypeDNA
Shipping ConditionDry Ice
Product LineHEX
Unit SizeEach
Contents & Storage
This reagent should be stored dessicated at -20°C.