2-Bromo-2-phenylacetophenone, 97%
2-Bromo-2-phenylacetophenone, 97%
2-Bromo-2-phenylacetophenone, 97%
Thermo Scientific Chemicals

2-Bromo-2-phenylacetophenone, 97%

CAS: 1484-50-0 | C14H11BrO | 275.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 403170250
Price (USD)
257.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
257.00
Each
Chemical Identifiers
CAS1484-50-0
IUPAC Name2-bromo-1,2-diphenylethan-1-one
Molecular FormulaC14H11BrO
InChI KeyZFFBIQMNKOJDJE-UHFFFAOYNA-N
SMILESBrC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point53°C to 58°C
Appearance (Color)White to beige to light brown
Infrared spectrumConforms
GC>=96.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only