3-Bromo-1-propanol, 92%
3-Bromo-1-propanol, 92%
3-Bromo-1-propanol, 92%
Thermo Scientific Chemicals

3-Bromo-1-propanol, 92%

CAS: 627-18-9 | C3H7BrO | 138.99 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
產品號碼 403200250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS627-18-9
IUPAC Name3-bromopropan-1-ol
Molecular FormulaC3H7BrO
InChI KeyRQFUZUMFPRMVDX-UHFFFAOYSA-N
SMILESOCCCBr
檢視更多
規格Specification Sheet規格表
Refractive index1.4850 to 1.4930 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow to light brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only