2-Bromopyrimidine, 97+%
2-Bromopyrimidine, 97+%
2-Bromopyrimidine, 97+%
Thermo Scientific Chemicals

2-Bromopyrimidine, 97+%

CAS: 4595-60-2 | C4H3BrN2 | 158.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4032300101 gGlass bottle
Catalog number 403230010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS4595-60-2
IUPAC Name2-bromopyrimidine
Molecular FormulaC4H3BrN2
InChI KeyPGFIHORVILKHIA-UHFFFAOYSA-N
SMILESBrC1=NC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point55°C to 59°C
Appearance (Color)Off-white
Infrared spectrumConforms
HPLC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only