Brucine sulfate heptahydrate, 98+%, ACS reagent
Brucine sulfate heptahydrate, 98+%, ACS reagent
Brucine sulfate heptahydrate, 98+%, ACS reagent
Thermo Scientific Chemicals

Brucine sulfate heptahydrate, 98+%, ACS reagent

CAS: 60583-39-3 | C46H68N4O19S | 1013.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
Packaging:
Glass bottle
Catalog number 403290100
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.40
Save 5.75
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
33.65
Special offer
Online exclusive
Ends: 28-Dec-2026
39.40
Save 5.75
Each
Chemical Identifiers
CAS60583-39-3
IUPAC Namebis((11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one) sulfuric acid heptahydrate
Molecular FormulaC46H68N4O19S
InChI KeyPPJIVFYMRNHHTJ-UMRIXEKQSA-N
SMILESO.O.O.O.O.O.O.OS(O)(=O)=O.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34.COC1=CC2=C(C=C1OC)C13CCN4CC5=CCO[C@H]6CC(=O)N2[C@H]1[C@H]6[C@H]5C[C@@H]34
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Residue after ignition=<0.1 %
Sensitivity(to nitrate) Passes test
Appearance (Form)Crystalline powder
Titration with HClO4>=98.0 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only