Butyl 4-hydroxybenzoate, 99+%
Butyl 4-hydroxybenzoate, 99+%
Butyl 4-hydroxybenzoate, 99+%
Thermo Scientific Chemicals

Butyl 4-hydroxybenzoate, 99+%

CAS: 94-26-8 | C11H14O3 | 194.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
Catalog number 403571000
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.80
Save 7.15
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Special offer
Online exclusive
Ends: 28-Dec-2026
48.80
Save 7.15
Each
Chemical Identifiers
CAS94-26-8
IUPAC Namebutyl 4-hydroxybenzoate
Molecular FormulaC11H14O3
InChI KeyQFOHBWFCKVYLES-UHFFFAOYSA-N
SMILESCCCCOC(=O)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Melting point68°C to 71°C
Infrared spectrumConforms
GC>=99.0 %
Appearance (Form)Crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only