1-chlorobutane, Sequenation Grade
1-chlorobutane, Sequenation Grade
1-chlorobutane, Sequenation Grade
Thermo Scientific Chemicals

1-chlorobutane, Sequenation Grade

CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4045500101 LGlass bottle
Catalog number 404550010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS109-69-3
IUPAC Name1-chlorobutane
Molecular FormulaC4H9Cl
InChI KeyVFWCMGCRMGJXDK-UHFFFAOYSA-N
SMILESCCCCCl
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=99.5 %
Residue after evaporation=<2 ppm
Appearance (Form)Liquid
UVat 220 nm A: =<1
UVat 300 nm A: =<0.005
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only