xMAP Sheath Fluid Plus
Invitrogen™
xMAP Sheath Fluid Plus
xMAP Sheath Fluid PLUS is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-basedRead more
Have Questions?
Catalog NumberQuantity
405002120 L
Catalog number 4050021
Price (USD)
92.25
Each
Quantity:
20 L
Price (USD)
92.25
Each
  • xMAP Sheath Fluid PLUS is the delivery medium that carries the sample to the optics component of Luminex xMAP technology-based instruments. It is needed to operate all Luminex 200, FLEXMAP 3D, xMAP INTELLIFLEX, and xMAP INTELLIFLEX DRSE systems.
  • This reformulated sheath product replaces the original xMAP Sheath Fluid (Cat. No. 4050015).
  • The new sheath formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
Specifications
CAS7732-18-5
PackagingContainer
Physical FormLiquid
Quantity20 L
Unit SizeEach
Frequently asked questions (FAQs)
What is the difference between xMAP Sheath Concentrate PLUS, RUO and xMAP Sheath Fluid Plus?
xMAP Sheath Concentrate PLUS, RUO (Cat. No. 4050023) is a 20X concentrated version of xMAP Sheath Fluid Plus (Cat. No. 4050021). It needs to be diluted before use.
xMAP Sheath Concentrate PLUS, RUO (Cat. No. 4050023) is a reformulation of xMAP Sheath Fluid Concentrate (Cat. No. 4050018). The formulation is updated from a single preservative to a mixture of three preservatives to allow for greater transit stability and to reduce the likelihood of future acquired microbial resistance.
Will there be any differences in performance of my instrument(s) when switching over to xMAP Sheath Fluid Plus from the original xMAP Sheath Fluid?
In-house testing has been done to confirm that assays deliver expected results when using xMAP Sheath Fluid Plus. White Paper is available upon request from Technical Support (techsupport@thermofisher.com).
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
Are there special instructions or considerations when switching over to xMAP Sheath Fluid Plus from the original xMAP Sheath Fluid?
Yes, there are instructions available for flushing the instrument and replacing the sheath fluid. Please contact Technical Support at techsupport@thermofisher.com for these instructions.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
What is the advantage of xMAP Sheath Fluid Plus over the original xMAP Sheath Fluid?
xMAP Sheath Fluid Plus (Cat. No. 4050021) replaces the original xMAP Sheath Fluid (Cat. No. 4050015). The new sheath formulation is updated from a single preservative to a mixture of three preservatives, specifically, sodium azide, sodium benzoate and diazolidinyl urea. This allows for greater transit stability and reduces the likelihood of future acquired microbial resistance.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.