Will there be any differences in performance of my instrument when switching over to MAGPIX Drive Fluid Plus from the original MAGPIX Drive Fluid?
In-house testing has been done to confirm that assays deliver expected results when using MAGPIX Drive Fluid Plus. White Paper is available upon request from Technical Support (techsupport@thermofisher.com).