Cyclohexanone, 99+%, ACS reagent
Cyclohexanone, 99+%, ACS reagent
Cyclohexanone, 99+%, ACS reagent
Thermo Scientific Chemicals

Cyclohexanone, 99+%, ACS reagent

CAS: 108-94-1 | C6H10O | 98.145 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Catalog number 406092500
Price (USD)
36.10
Each
Quantity:
250 mL
Request bulk or custom format
Price (USD)
36.10
Each
Chemical Identifiers
CAS108-94-1
IUPAC Namecyclohexanone
Molecular FormulaC6H10O
InChI KeyJHIVVAPYMSGYDF-UHFFFAOYSA-N
SMILESO=C1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<10 APHA
GC>=99.0 %
Water=<0.05 %
Residue after evaporation=<0.05 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only