Decyltrimethylammonium bromide, 99%
Decyltrimethylammonium bromide, 99%
Decyltrimethylammonium bromide, 99%
Thermo Scientific Chemicals

Decyltrimethylammonium bromide, 99%

CAS: 2082-84-0 | C13H30BrN | 280.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Catalog number 406230250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS2082-84-0
IUPAC Namedecyltrimethylazanium bromide
Molecular FormulaC13H30BrN
InChI KeyPLMFYJJFUUUCRZ-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCC[N+](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Titration Argentometric>=98.5 %
Infrared spectrumConforms
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only