trans-1,4-Diaminocyclohexane, 98+%
trans-1,4-Diaminocyclohexane, 98+%
trans-1,4-Diaminocyclohexane, 98+%
Thermo Scientific Chemicals

trans-1,4-Diaminocyclohexane, 98+%

CAS: 2615-25-0 | C6H14N2 | 114.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Plastic bottle
Catalog number 406361000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS2615-25-0
IUPAC Namecyclohexane-1,4-diamine
Molecular FormulaC6H14N2
InChI KeyVKIRRGRTJUUZHS-UHFFFAOYSA-N
SMILESNC1CCC(N)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point68°C to 73°C
Appearance (Color)White to yellow to brown
Appearance (Form)Crystals and/or chunks
Infrared spectrumConforms
GC>=98.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only