2,4-Dibromophenol, 99%
2,4-Dibromophenol, 99%
2,4-Dibromophenol, 99%
Thermo Scientific Chemicals

2,4-Dibromophenol, 99%

CAS: 615-58-7 | C6H4Br2O | 251.91 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4065100505 gGlass bottle
Catalog number 406510050
Price (USD)
22.65
Special offer
Online exclusive
Ends: 28-Dec-2026
26.20
Save 3.55
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
22.65
Special offer
Online exclusive
Ends: 28-Dec-2026
26.20
Save 3.55
Each
Chemical Identifiers
CAS615-58-7
IUPAC Name2,4-dibromophenol
Molecular FormulaC6H4Br2O
InChI KeyFAXWFCTVSHEODL-UHFFFAOYSA-N
SMILESOC1=CC=C(Br)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point36°C to 39°C
GC>=98.5 %
Infrared spectrumConforms
Appearance (Form)chunks
Appearance (Color)White to light yellow to beige
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only