1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.
Thermo Scientific Chemicals

1,2-Dichloroethane, 99+%, ACS reagent, meets the requirements of Reag. Ph.Eur.

CAS: 107-06-2 | C2H4Cl2 | 98.95 g/mol
数量:
500 mL
1 L
2.5 L
4 L
パッケージ:
すべて表示
Glass Bottle
製品番号(カタログ番号) 406825000
価格(JPY)
-
数量:
500 mL
パッケージ:
Glass Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS107-06-2
IUPAC Name1,2-dichloroethane
Molecular FormulaC2H4Cl2
InChI KeyWSLDOOZREJYCGB-UHFFFAOYSA-N
SMILESClCCCl
さらに表示
GC>=99.0 %
Residue after evaporation=<0.002 %
Appearance (Form)Clear liquid
Density(20/20°C) about 1.25
Color scale=<10 APHA
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only