Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™
Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™
Thermo Scientific Chemicals

Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™

Dichloromethane, 99.9%, for biochemistry, stabilized with approx. 50 ppm amylene, AcroSeal™. | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 406910025
Price (USD)
-
Quantity:
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
GC>=99.85 %
Alkalinity=<0.0002 meq/g
UVat 235 nm A: =<1.0
Residue after evaporation=<0.001 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only