Dichloromethane, 99.5%, for spectroscopy ACS, stabilized with amylene
Dichloromethane, 99.5%, for spectroscopy ACS, stabilized with amylene
Dichloromethane, 99.5%, for spectroscopy ACS, stabilized with amylene
Thermo Scientific Chemicals

Dichloromethane, 99.5%, for spectroscopy ACS, stabilized with amylene

CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass Bottle
Catalog number 406930025
Price (EUR)
175,00
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
175,00
Each
Chemical Identifiers
CAS75-09-2
IUPAC Namedichloromethane
Molecular FormulaCH2Cl2
InChI KeyYMWUJEATGCHHMB-UHFFFAOYSA-N
SMILESClCCl
View more
SpecificationsSpecification SheetSpecification Sheet
Titrable acid=<0.0003 meq/g
Appearance (Color)Clear
UVat 260 nm A: =<0.04
Color scale=<10 APHA
Residue after evaporation=<0.002 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
RUO – Research Use Only