Didecyl phthalate, 95%
Didecyl phthalate, 95%
Didecyl phthalate, 95%
Thermo Scientific Chemicals

Didecyl phthalate, 95%

CAS: 84-77-5 | C28H46O4 | 446.67 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 407100250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS84-77-5
IUPAC Name1,2-bis(decyl) benzene-1,2-dicarboxylate
Molecular FormulaC28H46O4
InChI KeyPGIBJVOPLXHHGS-UHFFFAOYSA-N
SMILESCCCCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Viscous liquid
GC>=94 %
Appearance (Color)Clear to hazy yellow
Infrared spectrumConforms
Refractive index1.4780 to 1.4840 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only