Diethyl oxalacetate sodium salt, 95%, pract.
Diethyl oxalacetate sodium salt, 95%, pract.
Diethyl oxalacetate sodium salt, 95%, pract.
Thermo Scientific Chemicals

Diethyl oxalacetate sodium salt, 95%, pract.

CAS: 40876-98-0 | C8H11NaO5 | 210.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 407371000
Price (USD)
-
Quantity:
100 g
Request bulk or custom format
Chemical Identifiers
CAS40876-98-0
IUPAC Namesodium 1,4-diethoxy-1,3,4-trioxobutan-2-ide
Molecular FormulaC8H11NaO5
InChI KeyJPTKZRPOIUYFTM-UHFFFAOYSA-N
SMILES[Na+].CCOC(=O)[CH-]C(=O)C(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)Beige to light orange
Infrared spectrumConforms
Titration with HClO4>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only