N,N-Diethyl-p-phenylenediamine, 98%
N,N-Diethyl-p-phenylenediamine, 98%
N,N-Diethyl-p-phenylenediamine, 98%
Thermo Scientific Chemicals

N,N-Diethyl-p-phenylenediamine, 98%

CAS: 93-05-0 | C10H16N2 | 164.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 407390050
Price (USD)
22.65
Special offer
Online exclusive
Ends: 28-Dec-2026
26.20
Save 3.55
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
22.65
Special offer
Online exclusive
Ends: 28-Dec-2026
26.20
Save 3.55
Each
Chemical Identifiers
CAS93-05-0
IUPAC NameN1,N1-diethylbenzene-1,4-diamine
Molecular FormulaC10H16N2
InChI KeyQNGVNLMMEQUVQK-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Color)Clear yellow-brown to dark brown to black
Infrared spectrumConforms
Appearance (Form)Liquid or low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only