Diethyl phosphate, 96%
Diethyl phosphate, 96%
Diethyl phosphate, 96%
Thermo Scientific Chemicals

Diethyl phosphate, 96%

CAS: 598-02-7 | C4H11O4P | 154.1 g/mol
Have Questions?
Catalog NumberQuantity
4074200505 g
Catalog number 407420050
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
111.65
Online exclusive
124.00
Save 12.35
Each
Chemical Identifiers
CAS598-02-7
IUPAC Namediethoxyphosphinic acid
Molecular FormulaC4H11O4P
InChI KeyUCQFCFPECQILOL-UHFFFAOYSA-N
SMILESCCOP(O)(=O)OCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with NaOH95 to 105 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only