Diisopropanolamine, 99%
Diisopropanolamine, 99%
Diisopropanolamine, 99%
Thermo Scientific Chemicals

Diisopropanolamine, 99%

CAS: 110-97-4 | C6H15NO2 | 133.19 g/mol
Have Questions?
Catalog NumberQuantityPackaging
407805000500 gGlass bottle
Catalog number 407805000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS110-97-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Titration with HClO4>=98.5 %
Appearance (Form)Low melting mass
Refractive index1.4615 to 1.4635 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only