2,6-Dimethyl-3,5-heptanedione, 99%
2,6-Dimethyl-3,5-heptanedione, 99%
2,6-Dimethyl-3,5-heptanedione, 99%
Thermo Scientific Chemicals

2,6-Dimethyl-3,5-heptanedione, 99%

CAS: 18362-64-6 | C9H16O2 | 156.225 g/mol
Have Questions?
Catalog NumberQuantityPackaging
4083600505 gGlass Bottle
Catalog number 408360050
Price (USD)
116.65
Special offer
Online exclusive
Ends: 28-Dec-2026
137.00
Save 20.35
Each
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
116.65
Special offer
Online exclusive
Ends: 28-Dec-2026
137.00
Save 20.35
Each
Chemical Identifiers
CAS18362-64-6
IUPAC Name2,6-dimethylheptane-3,5-dione
Molecular FormulaC9H16O2
InChI KeyCEGGECULKVTYMM-UHFFFAOYSA-N
SMILESCC(C)C(=O)CC(=O)C(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Form)Liquid
Refractive index1.4560 to 1.4600 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only