N,N-Dimethyloctadecylamine, 89%, tech.
N,N-Dimethyloctadecylamine, 89%, tech.
N,N-Dimethyloctadecylamine, 89%, tech.
Thermo Scientific Chemicals

N,N-Dimethyloctadecylamine, 89%, tech.

CAS: 124-28-7 | C20H44ClN | 334.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 408430010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS124-28-7
IUPAC Namedimethyl(octadecyl)azanium chloride
Molecular FormulaC20H44ClN
InChI KeyPFKRTWCFCOUBHS-UHFFFAOYSA-N
SMILES[Cl-].CCCCCCCCCCCCCCCCCC[NH+](C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid after melting
Infrared spectrumConforms
Titration>=95 % (Tertiary amine (MW 297))
GC>=84 % (C18)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only