2,2-Dimethyl-1,3-propanediol, 99%
2,2-Dimethyl-1,3-propanediol, 99%
2,2-Dimethyl-1,3-propanediol, 99%
Thermo Scientific Chemicals

2,2-Dimethyl-1,3-propanediol, 99%

CAS: 126-30-7 | C5H12O2 | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 408550250
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.20
Save 5.55
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
34.65
Special offer
Online exclusive
Ends: 28-Dec-2026
40.20
Save 5.55
Each
Chemical Identifiers
CAS126-30-7
IUPAC Name2,2-dimethylpropane-1,3-diol
Molecular FormulaC5H12O2
InChI KeySLCVBVWXLSEKPL-UHFFFAOYSA-N
SMILESCC(C)(CO)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point123°C to 127°C
Acid value=<0.1 mg KOH/g
Infrared spectrumConforms
GC>=98.5 %
Appearance (Color)White
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only