1,5-Diphenyl-1,5-pentanedione, 99%
1,5-Diphenyl-1,5-pentanedione, 99%
1,5-Diphenyl-1,5-pentanedione, 99%
Thermo Scientific Chemicals

1,5-Diphenyl-1,5-pentanedione, 99%

CAS: 6263-83-8 | C17H16O2 | 252.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40901010010 gGlass bottle
Catalog number 409010100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6263-83-8
IUPAC Name1,5-diphenylpentane-1,5-dione
Molecular FormulaC17H16O2
InChI KeyYOLLTWVIOASMFW-UHFFFAOYSA-N
SMILESO=C(CCCC(=O)C1=CC=CC=C1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.5 %
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point64°C to 67°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only