Diphenylphosphoryl azide, 98%
Diphenylphosphoryl azide, 98%
Diphenylphosphoryl azide, 98%
Thermo Scientific Chemicals

Diphenylphosphoryl azide, 98%

CAS: 26386-88-9 | C12H10N3O3P | 275.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 409045000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS26386-88-9
IUPAC Name{[azido(phenoxy)phosphoryl]oxy}benzene
Molecular FormulaC12H10N3O3P
InChI KeySORGEQQSQGNZFI-UHFFFAOYSA-N
SMILES[N-]=[N+]=NP(=O)(OC1=CC=CC=C1)OC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5500 to 1.5540 (20°C, 589 nm)
Appearance (Color)Clear yellow to amber
GC>=97.5 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, For Azidation/Diazo transfer
RUO – Research Use Only