DL-1,4-Dithiothreitol, 98%, Pure
DL-1,4-Dithiothreitol, 98%, Pure
DL-1,4-Dithiothreitol, 98%, Pure
Thermo Scientific Chemicals

DL-1,4-Dithiothreitol, 98%, Pure

CAS: 3483-12-3 | C4H10O2S2 | 154.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 409190250
Price (EUR)
417,65
Online exclusive
464,00
Save 46,35 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
417,65
Online exclusive
464,00
Save 46,35 (10%)
Each
Chemical Identifiers
CAS3483-12-3
IUPAC Name(2R,3R)-1,4-disulfanylbutane-2,3-diol
Molecular FormulaC4H10O2S2
InChI KeyVHJLVAABSRFDPM-IMJSIDKUSA-N
SMILESO[C@@H](CS)[C@@H](O)CS
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Melting point38°C to 43°C
Appearance (Form)Powder or beads
Titration Iodimetric97.5 to 102.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only