Dodecylamine hydrochloride, 99%
Dodecylamine hydrochloride, 99%
Dodecylamine hydrochloride, 99%
Thermo Scientific Chemicals

Dodecylamine hydrochloride, 99%

CAS: 929-73-7 | C12H28ClN | 221.81 g/mol
Have Questions?
Catalog NumberQuantityPackaging
40925010010 gGlass bottle
Catalog number 409250100
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.00
Save 9.35
Each
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
53.65
Special offer
Online exclusive
Ends: 28-Dec-2026
63.00
Save 9.35
Each
Chemical Identifiers
CAS929-73-7
IUPAC Namedodecan-1-aminium chloride
Molecular FormulaC12H28ClN
InChI KeyTWFQJFPTTMIETC-UHFFFAOYSA-N
SMILES[Cl-].CCCCCCCCCCCC[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only