Erythrosin B, pure, certified - FAQs
Erythrosin B, pure, certified - FAQs
View additional product information for Erythrosin B, pure, certified - FAQs (409450250, 409451000)
1 product FAQs found
What is the alternative to TrypanBlue for using it with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000?
The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).