Erythrosin B, pure, certified
Erythrosin B, pure, certified
Erythrosin B, pure, certified
Thermo Scientific Chemicals

Erythrosin B, pure, certified

CAS: 16423-68-0 | C20H6I4Na2O5 | 879.86 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 409451000
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
148.65
Online Exclusive
165.00
Save 16.35 (10%)
Each
Chemical Identifiers
CAS16423-68-0
IUPAC Namedisodium 2-(2,4,5,7-tetraiodo-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H6I4Na2O5
InChI KeyIINNWAYUJNWZRM-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=C(C=CC=C1)C1=C2C=C(I)C(=O)C(I)=C2OC2=C(I)C([O-])=C(I)C=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Red to brown
Infrared spectrumConforms
UV Lambda max524 to 527 nm
Appearance (Form)Powder
Dye content>=80 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only

Frequently asked questions (FAQs)

What is the alternative to TrypanBlue for using it with the Countess 3 FL Automated Cell Counter (Cat. No. AMQAF2000?

The suitable alternative would be Erythrosin B (Cat. No. 409451000, A14180.36, 189460250 and J62619.22).

Find additional tips, troubleshooting help, and resources within our Cell Analysis Support Center.