CD68 Monoclonal Antibody (815CU17), eFluor™ 570, eBioscience™ - FAQs
CD68 Monoclonal Antibody (815CU17), eFluor™ 570, eBioscience™ - FAQs
View additional product information for CD68 Monoclonal Antibody (815CU17), eFluor™ 570, eBioscience™ - FAQs (41-0687-82)