CD4 Monoclonal Antibody (N1UG0), eFluor™ 570, eBioscience™ - FAQs
CD4 Monoclonal Antibody (N1UG0), eFluor™ 570, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (N1UG0), eFluor™ 570, eBioscience™ - FAQs (41-2444-80, 41-2444-82)