CD323 (JAM3) Monoclonal Antibody (SHM33), eFluor™ 570, eBioscience™ - FAQs
CD323 (JAM3) Monoclonal Antibody (SHM33), eFluor™ 570, eBioscience™ - FAQs
View additional product information for CD323 (JAM3) Monoclonal Antibody (SHM33), eFluor™ 570, eBioscience™ - FAQs (41-3239-82)