Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs (41-4015-80, 41-4015-82)