Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 570, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), eFluor™ 570, eBioscience™ - FAQs (41-4031-80)