Rat anti-Mouse IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
Rat anti-Mouse IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG2a Secondary Antibody, eFluor™ 570, eBioscience™ - FAQs (41-4210-80, 41-4210-82)